Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skiv2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021337.2 | NP_067312.2 | 4 - 5 | 405 | 70 | |
GenBank mRNA | AK029763.1 | - | 4 - 5 | 394 | 70 | |
AK087810.1 | - | 4 - 5 | 400 | 70 | ||
AK139051.1 | - | 4 - 5 | 412 | 70 | ||
AK159854.1 | - | 4 - 5 | 406 | 70 | ||
AK169864.1 | - | 4 - 5 | 376 | 70 | ||
BC062925.1 | - | 4 - 5 | 393 | 70 | ||
BC065999.1 | - | 4 - 5 | 377 | 5686586 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |