Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252447.1 | NP_001239376.1 | 2 - 3 | 479 | 71 | |
NM_027260.3 | NP_081536.2 | 2 - 3 | 479 | 71 | ||
GenBank mRNA | AF513620.1 | - | 1 - 2 | 143 | 71 | |
AK012664.1 | - | 2 - 3 | 171 | 71 | ||
AK053297.1 | - | 2 - 3 | 182 | 71 | ||
AK089825.1 | - | 2 - 3 | 467 | 71 | ||
AK154485.1 | - | 2 - 3 | 192 | 71 | ||
BC013520.1 | - | 1 - 2 | 153 | 4236402 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |