Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252447.1 | NP_001239376.1 | 6 - 7 | 793 | 86 | |
NM_027260.3 | NP_081536.2 | 6 - 7 | 793 | 86 | ||
GenBank mRNA | AF513620.1 | - | 5 - 6 | 457 | 86 | |
AK012664.1 | - | 6 - 7 | 485 | 86 | ||
AK154485.1 | - | 6 - 7 | 506 | 86 | ||
BC013520.1 | - | 5 - 6 | 467 | 4236402 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |