Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194262.2 | NP_919238.1 | 4 - 5 | 485 | 85 | |
NM_198122.2 | NP_937755.1 | 4 - 5 | 485 | 85 | ||
GenBank mRNA | AK031062.1 | - | 4 - 5 | 468 | 85 | |
AK041414.1 | - | 4 - 5 | 482 | 85 | ||
AK136105.1 | - | 4 - 5 | 368 | 85 | ||
AK161669.1 | - | 4 - 5 | 471 | 85 | ||
BC137783.1 | - | 3 - 4 | 247 | 8860802 | 85 | |
BY720039.1 | - | 4 - 5 | 368 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |