Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011203.2 | NP_035333.2 | 14 - 15 | 2112 | 67 | |
GenBank mRNA | AK154107.1 | - | 14 - 15 | 2204 | 67 | |
BC051980.1 | - | 14 - 15 | 2112 | 6402545 | 67 | |
X63440.1 | - | 14 - 15 | 2107 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |