Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252292.1 | NP_001239221.1 | 4 - 5 | 619 | 83 | |
NM_001252293.1 | NP_001239222.1 | 3 - 4 | 340 | 83 | ||
NM_008590.2 | NP_032616.1 | 3 - 4 | 518 | 83 | ||
GenBank mRNA | AF482999.1 | - | 4 - 5 | 340 | 83 | |
AK031718.1 | - | 3 - 4 | 517 | 83 | ||
AK032881.1 | - | 3 - 4 | 517 | 83 | ||
AK034949.1 | - | 3 - 4 | 516 | 83 | ||
AK146457.1 | - | 4 - 5 | 619 | 83 | ||
AK168714.1 | - | 3 - 4 | 516 | 83 | ||
AK168732.1 | - | 3 - 4 | 516 | 83 | ||
AK168743.1 | - | 3 - 4 | 516 | 83 | ||
AK168995.1 | - | 3 - 4 | 516 | 83 | ||
AK169095.1 | - | 3 - 4 | 516 | 83 | ||
AK169266.1 | - | 3 - 4 | 516 | 83 | ||
BC006639.1 | - | 3 - 4 | 501 | 3482928 | 83 | |
CF916557.1 | - | 3 - 4 | 332 | 83 | ||
D16262.1 | - | 3 - 4 | 505 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |