Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201352.2 | NP_958740.2 | 14 - 15 | 2214 | 72 | |
GenBank mRNA | AK141189.1 | - | 14 - 15 | 2247 | 72 | |
AK154771.1 | - | 14 - 15 | 2351 | 72 | ||
BC024955.1 | - | 13 - 14 | 1500 | 4953174 | 72 | |
BC026428.1 | - | 10 - 11 | 1147 | 4224574 | 72 | |
BC082585.1 | - | 14 - 15 | 2214 | 30608279 | 72 | |
BC119442.1 | - | 5 - 6 | 874 | 72 | ||
BC119443.1 | - | 5 - 6 | 874 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |