Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029742.2 | NP_084018.1 | 2 - 3 | 327 | 94 | |
GenBank mRNA | AK085130.1 | - | 2 - 3 | 285 | 94 | |
BC060132.1 | - | 2 - 3 | 327 | 6409751 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |