Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021887.2 | NP_068687.1 | 7 - 8 | 1220 | 115 | |
GenBank mRNA | AB049137.1 | - | 7 - 8 | 1112 | 115 | |
AF254068.1 | - | 7 - 8 | 924 | 115 | ||
AF269134.1 | - | 6 - 7 | 782 | 115 | ||
AF279436.1 | - | 7 - 8 | 1172 | 115 | ||
AF477982.1 | - | 6 - 7 | 798 | 115 | ||
AF477983.1 | - | 6 - 7 | 798 | 115 | ||
AF477984.1 | - | 6 - 7 | 798 | 115 | ||
AF477985.1 | - | 6 - 7 | 798 | 115 | ||
AF477986.1 | - | 6 - 7 | 798 | 115 | ||
AK150824.1 | - | 7 - 8 | 1181 | 115 | ||
AK171826.1 | - | 7 - 8 | 1159 | 115 | ||
AK172032.1 | - | 7 - 8 | 1192 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |