Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039092.3 | NP_001034181.1 | 7 - 8 | 865 | 60 | |
NM_001039093.1 | NP_001034182.1 | 7 - 8 | 865 | 60 | ||
NM_153080.2 | NP_694720.2 | 7 - 8 | 865 | 60 | ||
GenBank mRNA | AF467887.1 | - | 7 - 8 | 845 | 60 | |
AK043095.1 | - | 7 - 8 | 845 | 60 | ||
AK054531.1 | - | 7 - 8 | 833 | 60 | ||
AK166040.1 | - | 7 - 8 | 865 | 60 | ||
AK189087.1 | - | 2 - 3 | 152 | 60 | ||
BC062947.1 | - | 7 - 8 | 829 | 30536491 | 60 | |
BC127266.1 | - | 7 - 8 | 831 | 30357295 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |