Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029775.2 | NP_084051.1 | 3 - 4 | 470 | 93 | |
GenBank mRNA | AK013933.1 | - | 3 - 4 | 488 | 93 | |
AK014737.1 | - | 3 - 4 | 512 | 93 | ||
AK134862.1 | - | 517 | 93 | |||
BC020048.1 | - | 3 - 4 | 506 | 93 | ||
BC020089.1 | - | 3 - 4 | 470 | 4013755 | 93 | |
BC049641.1 | - | 659 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |