Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003949.3 | NP_001003949.2 | 5 - 6 | 1014 | 58 | |
GenBank mRNA | AY096036.1 | - | 5 - 6 | 724 | 58 | |
BC005494.1 | - | 4 - 5 | 537 | 58 | ||
BC052787.1 | - | 4 - 5 | 556 | 58 | ||
BC080302.1 | - | 74 | 58 | |||
BC100297.1 | - | 2 - 3 | 151 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |