Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019553.2 | NP_062426.2 | 5 - 6 | 1211 | 113 | |
GenBank mRNA | AF159131.3 | - | 5 - 6 | 1172 | 113 | |
AK010281.1 | - | 5 - 6 | 1091 | 113 | ||
AK145578.1 | - | 5 - 6 | 1195 | 113 | ||
AK160095.1 | - | 5 - 6 | 1185 | 113 | ||
BC030895.1 | - | 4 - 5 | 643 | 113 | ||
BC043655.1 | - | 5 - 6 | 1183 | 5363948 | 113 | |
BC059237.1 | - | 5 - 6 | 1171 | 6412782 | 113 | |
BC060220.1 | - | 5 - 6 | 1163 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |