Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205367.1 | NP_001192296.1 | 6 - 7 | 832 | 142 | |
NM_009859.4 | NP_033989.2 | 7 - 8 | 835 | 142 | ||
GenBank mRNA | AK088997.1 | - | 7 - 8 | 622 | 142 | |
AK144127.1 | - | 7 - 8 | 624 | 142 | ||
AK220447.1 | - | 6 - 7 | 605 | 142 | ||
BC058587.1 | - | 6 - 7 | 636 | 6529169 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |