Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172506.2 | NP_766094.1 | 3 - 4 | 886 | 83 | |
GenBank mRNA | AF388037.1 | - | 2 - 3 | 342 | 83 | |
AK028770.1 | - | 3 - 4 | 907 | 83 | ||
AK028889.1 | - | 3 - 4 | 887 | 83 | ||
AK043379.1 | - | 3 - 4 | 896 | 83 | ||
BC062892.1 | - | 3 - 4 | 866 | 6418202 | 83 | |
BC078631.1 | - | 3 - 4 | 833 | 6515418 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |