Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 1 - 2 | 253 | 132 | |
GenBank mRNA | AK159194.1 | - | 1 - 2 | 254 | 132 | |
AK159474.1 | - | 1 - 2 | 186 | 132 | ||
AK170262.1 | - | 1 - 2 | 239 | 132 | ||
AK171304.1 | - | 1 - 2 | 239 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |