Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025687.3 | NP_079963.1 | 2 - 3 | 111 | 109 | |
GenBank mRNA | AF285582.1 | - | 2 - 3 | 169 | 109 | |
AK006285.1 | - | 2 - 3 | 155 | 109 | ||
AK014896.1 | - | 2 - 3 | 168 | 109 | ||
AK017776.1 | - | 2 - 3 | 167 | 109 | ||
BC048458.1 | - | 2 - 3 | 111 | 6775116 | 109 | |
BC061081.1 | - | 2 - 3 | 166 | 6773183 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |