Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183408.3 | NP_899668.1 | 4 - 5 | 816 | 87 | |
GenBank mRNA | AK153657.1 | - | 4 - 5 | 820 | 87 | |
AK160843.1 | - | 3 - 4 | 250 | 87 | ||
BC027224.1 | - | 4 - 5 | 512 | 87 | ||
BC044786.2 | - | 3 - 4 | 1378 | 87 | ||
BC139405.1 | - | 4 - 5 | 656 | 8862427 | 87 | |
BC139408.1 | - | 4 - 5 | 656 | 8862430 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |