Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027480.3 | NP_081756.4 | 14 - 15 | 1381 | 116 | |
GenBank mRNA | AK043638.1 | - | 12 - 13 | 2463 | 116 | |
AK173316.1 | - | 12 - 13 | 2347 | 116 | ||
BC048236.1 | - | 13 - 14 | 1286 | 6313999 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |