Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007630.2 | NP_031656.2 | 2 - 3 | 264 | 91 | |
GenBank mRNA | AK013312.1 | - | 2 - 3 | 233 | 91 | |
AK076122.1 | - | 2 - 3 | 258 | 91 | ||
BC008247.1 | - | 2 - 3 | 180 | 3156950 | 91 | |
BY300844.1 | - | 2 - 3 | 267 | 91 | ||
CT010286.1 | - | 2 - 3 | 150 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |