Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081490.2 | NP_001074959.1 | 3 - 4 | 309 | 57 | |
NM_023605.2 | NP_076094.2 | 3 - 4 | 611 | 57 | ||
GenBank mRNA | AB041585.1 | - | 3 - 4 | 580 | 57 | |
AK018482.1 | - | 3 - 4 | 562 | 57 | ||
AK077607.1 | - | 3 - 4 | 272 | 57 | ||
BC020074.1 | - | 3 - 4 | 582 | 3710611 | 57 | |
DV657928.1 | - | 3 - 4 | 297 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |