Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009532.4 | NP_033558.3 | 9 - 10 | 1245 | 59 | |
GenBank mRNA | AK168334.1 | - | 9 - 10 | 1195 | 59 | |
AK168727.1 | - | 9 - 10 | 1199 | 59 | ||
BC055900.1 | - | 9 - 10 | 1221 | 4207343 | 59 | |
BC085281.1 | - | 11 - 12 | 1627 | 30472386 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |