Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027829.3 | NP_082105.3 | 6 - 7 | 577 | 67 | |
GenBank mRNA | BF682661.1 | - | 6 - 7 | 511 | 67 | |
CB055593.1 | - | 5 - 6 | 577 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |