Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC028493.1 | - | 2 - 3 | 328 | 80 | |
BC038517.1 | - | 3 - 4 | 514 | 80 | ||
BC052783.1 | - | 3 - 4 | 472 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |