Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110265.1 | NP_001103735.1 | 9 - 10 | 1054 | 92 | |
NM_001284272.1 | NP_001271201.1 | 9 - 10 | 1054 | 92 | ||
NM_009445.2 | NP_033471.2 | 9 - 10 | 1054 | 92 | ||
GenBank mRNA | AK041487.1 | - | 9 - 10 | 1025 | 92 | |
AK049200.1 | - | 9 - 10 | 1049 | 92 | ||
AK164093.1 | - | 9 - 10 | 1053 | 92 | ||
AX775731.1 | - | 9 - 10 | 1020 | 92 | ||
BC058851.1 | - | 9 - 10 | 1040 | 30023533 | 92 | |
DV648812.1 | - | 6 - 7 | 540 | 92 | ||
M86377.1 | - | 10 - 11 | 1080 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |