Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009359.4 | NP_033385.2 | 4 - 5 | 259 | 64 | |
GenBank mRNA | AB000619.1 | - | 5 - 6 | 507 | 64 | |
AK008505.1 | - | 4 - 5 | 257 | 64 | ||
AK012189.1 | - | 4 - 5 | 258 | 64 | ||
AK018568.1 | - | 257 | 64 | |||
AK141174.1 | - | 3 - 4 | 369 | 64 | ||
BC141033.1 | - | 4 - 5 | 401 | 9055645 | 64 | |
BC145184.1 | - | 4 - 5 | 401 | 64 | ||
BC145185.1 | - | 4 - 5 | 401 | 9053736 | 64 | |
BC145186.1 | - | 4 - 5 | 401 | 9053737 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |