Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173777.3 | NP_776138.2 | 3 - 4 | 398 | 61 | |
GenBank mRNA | AK037199.1 | - | 3 - 4 | 413 | 61 | |
BC115981.1 | - | 3 - 4 | 398 | 40106192 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |