Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114328.2 | NP_001107800.1 | 4 - 5 | 437 | 138 | |
NM_001286544.1 | NP_001273473.1 | 4 - 5 | 437 | 138 | ||
NM_001286545.1 | NP_001273474.1 | 4 - 5 | 437 | 138 | ||
NM_001286546.1 | NP_001273475.1 | 4 - 5 | 437 | 138 | ||
NM_028181.5 | NP_082457.3 | 4 - 5 | 437 | 138 | ||
GenBank mRNA | AK007973.1 | - | 4 - 5 | 334 | 138 | |
AK032645.1 | - | 4 - 5 | 323 | 138 | ||
AK076325.2 | - | 4 - 5 | 364 | 138 | ||
AK128894.1 | - | 4 - 5 | 363 | 138 | ||
AK134675.1 | - | 4 - 5 | 334 | 138 | ||
BC043049.1 | - | 4 - 5 | 336 | 5704185 | 138 | |
BC082602.1 | - | 4 - 5 | 338 | 30663407 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |