Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289656.1 | NP_001276585.1 | 16 - 17 | 2818 | 84 | |
NM_001289657.1 | NP_001276586.1 | 15 - 16 | 2792 | 84 | ||
NM_178630.4 | NP_848745.2 | 15 - 16 | 2803 | 84 | ||
GenBank mRNA | AK029793.1 | - | 16 - 17 | 2819 | 84 | |
AK029798.1 | - | 15 - 16 | 2376 | 84 | ||
DQ867031.1 | - | 15 - 16 | 2804 | 84 | ||
DQ867032.1 | - | 15 - 16 | 2804 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |