Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029669.4 | NP_083945.1 | 2 - 3 | 311 | 78 | |
GenBank mRNA | AK012521.1 | - | 2 - 3 | 264 | 78 | |
AK030483.1 | - | 2 - 3 | 289 | 78 | ||
BC026452.1 | - | 2 - 3 | 273 | 4460764 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |