Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010280.3 | NP_034410.3 | 5 - 6 | 1051 | 68 | |
GenBank mRNA | AB008833.1 | - | 5 - 6 | 929 | 68 | |
AF020305.1 | - | 5 - 6 | 884 | 68 | ||
AF036163.1 | - | 5 - 6 | 945 | 68 | ||
AF041842.1 | - | 5 - 6 | 934 | 68 | ||
AF051766.1 | - | 5 - 6 | 946 | 68 | ||
AK046542.1 | - | 5 - 6 | 978 | 68 | ||
BC066202.1 | - | 5 - 6 | 1051 | 6430904 | 68 | |
Y15110.1 | - | 5 - 6 | 965 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |