Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081256.1 | NP_001074725.1 | 4 - 5 | 710 | 117 | |
GenBank mRNA | AK129281.1 | - | 4 - 5 | 666 | 117 | |
AK147120.1 | - | 4 - 5 | 653 | 117 | ||
AK163281.1 | - | 5 - 6 | 706 | 117 | ||
BC151084.1 | - | 4 - 5 | 671 | 9087997 | 117 | |
BC172150.1 | - | 4 - 5 | 671 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |