Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019776.2 | NP_062750.2 | 11 - 12 | 1461 | 79 | |
GenBank mRNA | AB021491.1 | - | 11 - 12 | 1418 | 79 | |
AK034914.1 | - | 10 - 11 | 1330 | 79 | ||
AK088028.1 | - | 11 - 12 | 1420 | 79 | ||
AK133837.1 | - | 11 - 12 | 1467 | 79 | ||
AK159837.1 | - | 11 - 12 | 1426 | 79 | ||
AK161591.1 | - | 11 - 12 | 1415 | 79 | ||
AK162432.1 | - | 11 - 12 | 1402 | 79 | ||
AK167576.1 | - | 11 - 12 | 1451 | 79 | ||
BC007126.1 | - | 11 - 12 | 1408 | 3592548 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |