Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012042.3 | NP_036172.1 | 6 - 7 | 947 | 152 | |
GenBank mRNA | AF136441.1 | - | 5 - 6 | 802 | 152 | |
AF176910.1 | - | 6 - 7 | 836 | 152 | ||
AK147004.1 | - | 6 - 7 | 929 | 152 | ||
AK164274.1 | - | 6 - 7 | 914 | 152 | ||
BC004836.1 | - | 5 - 6 | 756 | 152 | ||
BC029260.1 | - | 6 - 7 | 947 | 2812400 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |