Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167757.1 | NP_001161229.1 | 4 - 5 | 811 | 121 | |
NM_029202.2 | NP_083478.2 | 4 - 5 | 808 | 121 | ||
GenBank mRNA | AK015948.1 | - | 4 - 5 | 752 | 121 | |
AK077080.1 | - | 4 - 5 | 780 | 121 | ||
BC049731.1 | - | 4 - 5 | 808 | 6771078 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |