Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600015349.mm.1 |
TC0600001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600015349.mm.1 |
NM_008174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015349.mm.1 |
NM_008174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015352.mm.1 |
TC0600001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600015352.mm.1 |
NM_008174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015352.mm.1 |
NM_008174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015347.mm.1 |
TC0600001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600015347.mm.1 |
NM_008174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015347.mm.1 |
NM_008174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015350.mm.1 |
TC0600001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600015350.mm.1 |
NM_008174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015350.mm.1 |
NM_008174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015351.mm.1 |
TC0600001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600015351.mm.1 |
NM_008174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015351.mm.1 |
NM_008174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015358.mm.1 |
TC0600001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600015358.mm.1 |
NM_008174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015358.mm.1 |
NM_008174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015348.mm.1 |
TC0600001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600015348.mm.1 |
NM_008174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600015348.mm.1 |
NM_008174.2 |
Refseq |