Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054073.2 | NP_473414.1 | 6 - 7 | 745 | 143 | |
GenBank mRNA | AF294348.1 | - | 6 - 7 | 713 | 143 | |
AK006265.1 | - | 6 - 7 | 746 | 143 | ||
BC125597.1 | - | 5 - 6 | 663 | 40130112 | 143 | |
BC125599.1 | - | 5 - 6 | 663 | 40130114 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |