Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010832.5 | NP_034962.2 | 1 - 2 | 169 | 72 | |
GenBank mRNA | AF117066.2 | - | 1 - 2 | 171 | 72 | |
AK146208.1 | - | 1 - 2 | 106 | 72 | ||
AK148898.1 | - | 1 - 2 | 148 | 72 | ||
BC010226.1 | - | 1 - 2 | 130 | 3501126 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |