Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289546.1 | NP_001276475.1 | 19 - 20 | 1946 | 87 | |
NM_175007.2 | NP_778172.1 | 19 - 20 | 1934 | 87 | ||
GenBank mRNA | AK132168.1 | - | 19 - 20 | 1947 | 87 | |
BC024817.1 | - | 9 - 10 | 915 | 87 | ||
BC043718.1 | - | 3 - 4 | 603 | 87 | ||
BC051425.1 | - | 5 - 6 | 3256 | 87 | ||
BC054718.1 | - | 19 - 20 | 1914 | 5686478 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |