Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081058.2 | NP_001074527.1 | 10 - 11 | 3346 | 84 | |
NM_027118.1 | NP_081394.1 | 10 - 11 | 3346 | 84 | ||
GenBank mRNA | AK035493.1 | - | 10 - 11 | 2017 | 84 | |
AK173265.1 | - | 10 - 11 | 3346 | 84 | ||
BC051093.1 | - | 3 - 4 | 230 | 84 | ||
BC076619.1 | - | 11 - 12 | 1801 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |