Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncald | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170868.1 | NP_001164339.1 | 1 - 2 | 163 | 79 | |
GenBank mRNA | AK167313.1 | - | 1 - 2 | 133 | 79 | |
BC011162.1 | - | 1 - 2 | 158 | 4191195 | 79 | |
BC026979.1 | - | 1 - 2 | 117 | 5365560 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |