Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008130.2 | NP_032156.2 | 12 - 13 | 2237 | 63 | |
GenBank mRNA | BC141135.1 | - | 12 - 13 | 1938 | 9055747 | 63 |
CA752350.1 | - | 1 - 2 | 95 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |