Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016762.2 | NP_058042.2 | 1 - 2 | 228 | 88 | |
GenBank mRNA | AK035102.1 | - | 1 - 2 | 230 | 88 | |
AK083151.1 | - | 1 - 2 | 225 | 88 | ||
AK138211.1 | - | 1 - 2 | 225 | 88 | ||
AK163818.1 | - | 1 - 2 | 187 | 88 | ||
BC005429.1 | - | 1 - 2 | 98 | 3492881 | 88 | |
BC092298.1 | - | 1 - 2 | 171 | 5063535 | 88 | |
BY146293.1 | - | 1 - 2 | 228 | 88 | ||
U69262.1 | - | 1 - 2 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |