Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020052.2 | NP_064436.2 | 4 - 5 | 609 | 70 | |
GenBank mRNA | AK142950.1 | - | 4 - 5 | 575 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |