Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177151.3 | NP_796125.2 | 11 - 12 | 1821 | 70 | |
GenBank mRNA | BC158052.1 | - | 11 - 12 | 1801 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |