Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203491.3 | NP_987076.2 | 2 - 3 | 427 | 61 | |
GenBank mRNA | BC019470.1 | - | 2 - 3 | 427 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Ion Channel Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |