Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026200.3 | NP_080476.2 | 1 - 2 | 764 | 73 | |
GenBank mRNA | AK012275.1 | - | 1 - 2 | 559 | 73 | |
AK034091.1 | - | 1 - 2 | 657 | 73 | ||
AK038450.1 | - | 1 - 2 | 751 | 73 | ||
AK076120.1 | - | 1 - 2 | 533 | 73 | ||
AK139482.1 | - | 1 - 2 | 600 | 73 | ||
BC117819.1 | - | 1 - 2 | 653 | 73 | ||
BC128477.1 | - | 1 - 2 | 653 | 40092817 | 73 | |
BC128478.1 | - | 1 - 2 | 653 | 40092823 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus 1700 Probe spans exons |