Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000004991.mm.1 |
NM_001081412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004991.mm.1 |
NM_001081412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004991.mm.1 |
TC1000000708.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004992.mm.1 |
NM_001081412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004992.mm.1 |
NM_001081412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004992.mm.1 |
TC1000000708.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004990.mm.1 |
NM_001081412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004990.mm.1 |
NM_001081412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004990.mm.1 |
TC1000000708.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004993.mm.1 |
NM_001081412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004993.mm.1 |
NM_001081412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004993.mm.1 |
TC1000000708.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004994.mm.1 |
NM_001081412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004994.mm.1 |
NM_001081412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004994.mm.1 |
TC1000000708.mm.1 |
MTA TC |