Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181816.2 | NP_861537.2 | 9 - 10 | 1194 | 76 | |
GenBank mRNA | AK077147.1 | - | 4 - 5 | 572 | 76 | |
BC096547.1 | - | 8 - 9 | 1087 | 30360184 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |