Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145884.1 | NP_001139356.1 | 10 - 11 | 1988 | 80 | |
NM_010580.2 | NP_034710.2 | 10 - 11 | 1988 | 80 | ||
GenBank mRNA | AF022110.1 | - | 10 - 11 | 1987 | 80 | |
AF043256.1 | - | 10 - 11 | 1893 | 80 | ||
AF043257.1 | - | 10 - 11 | 1893 | 80 | ||
AK087476.1 | - | 10 - 11 | 1735 | 80 | ||
AK140251.1 | - | 10 - 11 | 1900 | 80 | ||
AK143532.1 | - | 10 - 11 | 1923 | 80 | ||
AK187695.1 | - | 1 - 2 | 124 | 80 | ||
BC029170.1 | - | 5 - 6 | 835 | 80 | ||
BC046280.1 | - | 10 - 11 | 1897 | 6394641 | 80 | |
BC058246.1 | - | 10 - 11 | 1952 | 6313668 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |